missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BEX1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA524192

Product Code. 13218518

  • £487.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is predicted to react with non-human primate based on sequence homology.

Signaling adapter molecule involved in p75NTR/NGFR signaling. Plays a role in cell cycle progression and neuronal differentiation. Inhibits neuronal differentiation in response to nerve growth factor (NGF). May act as a link between the cell cycle and neurotrophic factor signaling, possibly by functioning as an upstream modulator of receptor signaling, coordinating biological responses to external signals with internal cellular states.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BEX1
Polyclonal
Unconjugated
BEX1
AA407839; AU018692; BEX1; BEX2; Bex4; brain expressed gene 1; brain expressed X-linked 1; brain expressed, X-linked 1; brain-expressed X-linked protein 1; Brain-expressed X-linked protein 1 homolog; EG2RVC; H HGR74-h; HBEX2; HGR74-h; ovarian granulosa cell 13.0 kDa protein hGR74; protein BEX1; reduced expression 3; reduced expression protein 3; Rex3; REX-3
Rabbit
Antigen affinity chromatography, Protein A
RUO
55859
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9HBH7
BEX1
KLH conjugated synthetic peptide between 63-90 amino acids from the central region of human BEX1.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.