missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BEX1/BEX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100404
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BEX1/2.
BEX1/BEX2 belongs to the brain expressed X-linked gene family. The encoded protein interacts with the transcription factor LIM domain only 2 in a DNA-binding complex that recognizes the E-box element and promotes transcription. BEX1/BEX2 has been found to be a tumor suppressor that is silenced in human glioma. In breast cancer cells, this gene product modulates apoptosis in response to estrogen and tamoxifen, and enhances the anti-proliferative effect of tamoxifen. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| BEX1/BEX2 | |
| Polyclonal | |
| Unconjugated | |
| BEX1 | |
| AA407839; AL024066; AU018692; BEX1; BEX2; Bex4; brain expressed gene 1; brain expressed X-linked 1; brain expressed X-linked 2; brain expressed, X-linked 1; brain-expressed X-linked protein 1; Brain-expressed X-linked protein 1 homolog; brain-expressed X-linked protein 2; Brain-expressed X-linked protein 2 homolog; DJ79P11.1; EG2RVC; expression gene 2 in rat visual cortex; hBex2; HGR74-h; ovarian granulosa cell 13.0 kDa protein hGR74; protein BEX1; Protein BEX2; protein hBex2; reduced expression 3; reduced expression protein 3; Rex3; REX-3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12069, 19716, 363498, 501625, 55859, 84707 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3MKQ1, Q3MKQ2, Q9BXY8, Q9HBH7, Q9R224, Q9WTZ8 | |
| BEX1, BEX2 | |
| A synthesized peptide derived from human BEX2(Accession Q9BXY8), corresponding to amino acid residues M1-R50. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction