missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bevacizumab Mouse, Biotin, UltraLys, Monoclonal Antibody, AcroBiosystems
Mouse Monoclonal Antibody
£352.00 - £1510.00
Specifications
| Antigen | Bevacizumab |
|---|---|
| Classification | Monoclonal |
| Conjugate | Biotin |
| Host Species | Hybridoma |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16477219
|
ACROBiosystems
BEB-BY13-25ug |
25 μg |
£352.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16467219
|
ACROBiosystems
BEB-BY13-200ug |
200 μg |
£1510.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bevacizumab Monoclonal antibody specifically detects Bevacizumab, recommended for PK/PD.Specifications
| Bevacizumab | |
| Biotin | |
| RUO | |
| -20°C |
| Monoclonal | |
| Hybridoma | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title