missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Bevacizumab

Product Code. 16492278
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492278 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492278 Supplier MedChemExpress Supplier No. HYP9906

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bevacizumab, a humanized IgG1 monoclonal antibody, specifically binds to all VEGF-A isoforms with high affinity.

Specifications

Antigen VEGF
Quantity 5 mg
Target Species Human
Content And Storage Please store the product under the recommended conditions in the Certificate of Analysis.
Product Type Inhibitory Antibodies
Shipping Condition Room temperature
CAS 216974-75-3
Purity or Quality Grade 97.7%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.