missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Bevacizumab
Specifications
Specifications
| Antigen | VEGF |
| Quantity | 1 mg |
| Target Species | Human |
| Content And Storage | Please store the product under the recommended conditions in the Certificate of Analysis. |
| Product Type | Inhibitory Antibodies |
| Shipping Condition | Room temperature |
| CAS | 216974-75-3 |
| Purity or Quality Grade | 97.7% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?