missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Betulonic Acid, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Betulonic acid |
| CAS | 4481-62-3 |
| Molecular Formula | C30 H46 O3 |
| Quantity | 100 mg |
| Synonym | 3-Oxo-lup-20(29)-en-28-oic acid, 3-Oxobetulinic acid |
| InChI Formula | InChI=1S/C30H46O3/c1-18(2)19-10-15-30(25(32)33)17-16-28(6)20(24(19)30)8-9-22-27(5)13-12-23(31)26(3,4)21(27)11-14-29(22,28)7/h19-22,24H,1,8-17H2,2-7H3,(H,32,33)/t19-,20+,21-,22+,24+,27-,28+,29+,30-/m0/s1 |
| SMILES | CC(=C)[C@@H]1CC[C@@]2(CC[C@]3(C)[C@H](CC[C@@H]4[C@@]5(C)CCC(=O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12)C(=O)O |
| IUPAC Name | (1R,3aS,5aR,5bR,7aR,11aR,11bR,13aR,13bR)-1-isopropenyl-5a,5b,8,8,11a-pentamethyl-9-oxo-2,3,4,5,6,7,7a,10,11,11b,12,13,13a,13b-tetradecahydro-1H-cyclopenta[a]chrysene-3a-carboxylic acid |
| Molecular Weight (g/mol) | 454.68 |
| Formula Weight | 454.3447 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction