missing translation for 'onlineSavingsMsg'
Learn More

Betulinic Acid, TRC

Product Code. 30011166
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011166 1 g 1g
30012281 100 mg 100mg
30009243 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011166 Supplier Toronto Research Chemicals Supplier No. TRCB3302501G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 472-15-1
Molecular Formula C30 H48 O3
Molecular Weight (g/mol) 456.7
Synonym (3β)-3-Hydroxy-lup-20(29)-en-28-oic acid, Lup-20(29)-en-28-oic acid, 3β-hydroxy- (8CI), Mairin (6CI,7CI), 3aH-Cyclopenta[a]chrysene, lup-20(29)-en-28-oic acid deriv., (+)-Betulinic acid, 3β-Hydroxylup-20(29)-en-28-oic acid, Betulic acid, Betulinic acid, Lupatic acid, NSC 113090, β-Betulinic acid
IUPAC Name (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta[a]chrysene-3a-carboxylic acid
SMILES CC(=C)[C@@H]1CC[C@@]2(CC[C@]3(C)[C@H](CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C30H48O3/c1-18(2)19-10-15-30(25(32)33)17-16-28(6)20(24(19)30)8-9-22-27(5)13-12-23(31)26(3,4)21(27)11-14-29(22,28)7/h19-24,31H,1,8-17H2,2-7H3,(H,32,33)/t19-,20+,21-,22+,23-,24+,27-,28+,29+,30-/m0/s1
Formula Weight 456.3603
Percent Purity >95%
Chemical Name or Material Betulinic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.