missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Betulinic acid
GREENER_CHOICE

Product Code. p-4816561
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18084452

Brand: Tocris Bioscience™ 3906/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antitumor and anti-HIV agent; activates NF-κB. Also TGR5 agonist

Betulinic acid is a natural triterpenoid that displays anti-HIV and antitumor activity. Induces the production of reactive oxygen species (ROS) and activates NF-κB. Also a GPBA receptor partial agonist (EC50 = 1.04 μM, efficacy 83%). Betulinic acid suppresses areobic glycolysis by regulating the Cav-1/NF-κB/c-Myc pathway in and inhibits growth in breast cancer cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 472-15-1
Molecular Formula C30H48O3

Specifications

Quantity 10 mg
Formula Weight 456.7
Purity >98%
Molecular Weight (g/mol) 456.711
Chemical Name or Material Betulinic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.