missing translation for 'onlineSavingsMsg'
Learn More

Betulinic acid, MedChemExpress

Product Code. 16420996
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16420996

Brand: Medchem Express HY10529

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Betulinic acid is a natural pentacyclic triterpenoid, acts as a eukaryotic topoisomerase I inhibitor, with an IC50 of 5 μM, and possesses anti-HIV, anti-malarial, anti-inflammatory and anti-tumor properties.

Betulinic acid is a natural pentacyclic triterpenoid, acts as a eukaryotic topoisomerase I inhibitor, with an IC50 of 5 μM, and possesses anti-HIV, anti-malarial, anti-inflammatory and anti-tumor properties.

Specifications

Chemical Name or Material Betulinic acid
CAS 472-15-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H48O3
Quantity 200 mg
Synonym Lupatic acid Betulic acid
Solubility Information DMSO : 35.71 mg/mL (78.19 mM; Need ultrasonic)
SMILES [H][C@@]12CC[C@@]3(C)[C@]4(C)CC[C@]5(C(O)=O)[C@@]([C@H](C(C)=C)CC5)([H])[C@@](CC[C@@]([H])3[C@@]1(C)CC[C@H](O)C2(C)C)4[H]
Molecular Weight (g/mol) 456.7
Formula Weight 456.7
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.