missing translation for 'onlineSavingsMsg'
Learn More

BETd-260, MedChemExpress

Product Code. 16493398
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493398 10 mg 10mg
16479926 50 mg 50mg
16456127 5 mg 5mg
16432756 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493398 Supplier Medchem Express Supplier No. HY101519
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BETd-260 (ZBC 260) is a PROTAC connected by ligands for Cereblon and BET, with as low as 30 pM against BRD4 protein in RS4;11 leukemia cell line. BETd-260 potently suppresses cell viability and robustly induces apoptosis in hepatocellular carcinoma (HCC) cells.

BETd-260 (ZBC 260) is a PROTAC connected by ligands for Cereblon and BET, with as low as 30 pM against BRD4 protein in RS4

Specifications

Chemical Name or Material BETd-260
CAS 2093388-62-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C43H46N10O6
Quantity 10 mg
Synonym ZBC 260
Solubility Information DMSO : 25 mg/mL (31.29 mM; Need ultrasonic)
SMILES CCN1N=C(C2CC2)C=C1NC3=NC(C(NCCCCCC4=CC=CC5=C4CN(C6C(NC(CC6)=O)=O)C5=O)=O)=NC7=C3C8=CC(OC)=C(C9=C(C)ON=C9C)C=C8N7
Molecular Weight (g/mol) 798.89
Formula Weight 798.89
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.