missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Betaxolol hydrochloride

Product Code. 16594214 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16594214 1 g 1g
16504224 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16594214 Supplier Thermo Scientific Chemicals Supplier No. 462440010
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63659-19-8
Molecular Formula C18H30ClNO3
Molecular Weight (g/mol) 343.89
InChI Key CHDPSNLJFOQTRK-UHFFFAOYNA-N
IUPAC Name hydrogen 1-{4-[2-(cyclopropylmethoxy)ethyl]phenoxy}-3-[(propan-2-yl)amino]propan-2-ol chloride
SMILES [H+].[Cl-].CC(C)NCC(O)COC1=CC=C(CCOCC2CC2)C=C1

Specifications

Melting Point 115.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: very soluble aq. soln.. Other solubilities: very soluble in methanol and ethanol, slightly soluble in acetone and acetonitrile, insoluble in ether
Formula Weight 343.89
Chemical Name or Material βxolol hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.