missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ betatrophin Monoclonal Antibody (D3)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA543939

 View more versions of this product

Product Code. 17634396

  • £178.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

betatrophin Monoclonal Antibody for Western Blot

The Betatrophin protein, also known as the hepatocellular carcinoma-associated protein TD26, is an adipocyte- and liver-enriched insulin targeted gene that is thought to play a role in lipid metabolism be regulating serum triglyceride levels. Betatrophin is closely related to the angiopoietin-like (ANGPTL) protein family. Betatrophin mRNA increases in the cold environment and after treatment with a high-fat diet, while its mRNA levels decrease in response to fasting, suggesting that it may play a role in mediating thermogenesis and energy homeostasis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

betatrophin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q6UXH0
ANGPTL8
Recombinant protein Angiopoietin Like Protein 8. The antigen corresponds to amino acid range 24-196 of the target protein.
20 μL
Primary
Human, Pig
Antibody
IgG1 κ
Western Blot
D3
Unconjugated
ANGPTL8
angiopoietin like 8; angiopoietin-like 8; angiopoietin-like protein 8; Angptl8; betatrophin; betatrophin variant 1; betatrophin variant 2; C19orf80; EG624219; Gm6484; hepatocellular carcinoma-associated gene TD26; hepatocellular carcinoma-associated protein TD26; hepatocellular carcinoma-associated protein TD26 homolog; lipasin; LOC100361444; predicted gene 6484; PRO1185; PVPA599; rCG31799-like; refeeding-induced fat and liver protein; Rifl; TD26; UNQ599/PRO1185
Mouse
Protein A/G
RUO
110259345, 55908
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.