missing translation for 'onlineSavingsMsg'
Learn More

(BetaR,DeltaR,1S,2S)-1,2-Dihydro-Beta,Delta,6-trihydroxy-2-methyl-1-naphthaleneheptanoic Acid Sodium Salt (Pravastatin Impurity), TRC

Product Code. 30388643
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388643 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388643 Supplier Toronto Research Chemicals Supplier No. TRCD44986010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3R,5R)-3,5-Dihydroxy-7-[(1S,2S)-6-hydroxy-2-methyl-1,2-dihydronaphthalen-1-yl]heptanoic Acid Sodium Salt
CAS 720666-74-0
Molecular Formula C18 H23 O5 . Na
Quantity 10 mg
Synonym 1-Naphthaleneheptanoic acid, 1,2-dihydro-beta,delta,6-trihydroxy-2-methyl-, sodium salt (1:1), (betaR,deltaR,1S,2S)-, 1-Naphthaleneheptanoic acid, 1,2-dihydro-beta,delta,6-trihydroxy-2-methyl-, monosodium salt, (betaR,deltaR,1S,2S)- (9CI), Imp. G (EP) as Sodium Salt: (3R,5R)-3,5-Dihydroxy-7-[(1S,2S)-6-hydroxy-2-methyl-1,2-dihydronaphthalen-1-yl]heptanoic Acid Sodium Salt, Pravastatin Impurity, R 195 Sodium Salt
InChI Formula InChI=1S/C18H24O5.Na/c1-11-2-3-12-8-13(19)5-7-17(12)16(11)6-4-14(20)9-15(21)10-18(22)23;/h2-3,5,7-8,11,14-16,19-21H,4,6,9-10H2,1H3,(H,22,23);/q;+1/p-1/t11-,14+,15+,16-;/m0./s1
SMILES [Na+].C[C@H]1C=Cc2cc(O)ccc2[C@H]1CC[C@@H](O)C[C@@H](O)CC(=O)[O-]
IUPAC Name sodium;(3R,5R)-3,5-dihydroxy-7-[(1S,2S)-6-hydroxy-2-methyl-1,2-dihydronaphthalen-1-yl]heptanoate
Molecular Weight (g/mol) 342.36
Formula Weight 342.1443
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.