missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betamethasone Valerate (Betamethasone 17-Valerate), LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Betamethasone Valerate |
| CAS | 2152-44-5 |
| Analyte or Component Names | Betamethasone Valerate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H37 F O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Betamethasone 17-Valerate, Betamethasone Valerate, Bedermin, Beta-Val, Betamethasone 17-valerate, Betamethasone valerate, Betnesol V, Betneval, Betnovate, Betnovateat, Betoid, Bettamousse, Bextasol, Celestan V, Celestane V, Celestoderm, Celestoderm V, Celeston valerate, Dermosol, Dermovaleas, Ecoval 70, Fuciderm, Hormezon, Rinderon V, Stanoval, Tokuderm, Valisone, (11beta,16beta)-9-Fluoro-11,21-dihydroxy-16-methyl-17-[(1-oxopentyl)oxy]pregna-1,4-diene-3,20-dione, 9-Fluoro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione 17-valerate, Valeric acid 17-ester with 9-fluoro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione, 9alpha-Fluoro-11beta,21-dihydroxy-16beta-methyl-17alpha-valeryloxypregna-1,4-diene-3,20-dione, 9alpha-Fluoro-16beta-methylprednisolone 17-valerate, Betamethasone 17alpha-valerate, beta-Methasone 17-valerate, CS valerate, Betamethasone-21-valerate |
| InChI Formula | InChI=1S/C27H37FO6/c1-5-6-7-23(33)34-27(22(32)15-29)16(2)12-20-19-9-8-17-13-18(30)10-11-24(17,3)26(19,28)21(31)14-25(20,27)4/h10-11,13,16,19-21,29,31H,5-9,12,14-15H2,1-4H3/t16-,19-,20-,21-,24-,25-,26-,27-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction