missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betamethasone 17-Valerate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Betamethasone 17-Valerate
Specifications
Specifications
| Chemical Name or Material | Betamethasone Valerate |
| CAS | 2152-44-5 |
| Analyte or Component Names | Betamethasone Valerate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H37 F O6 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Betamethasone 17-Valerate, Betamethasone Valerate, Bedermin, Beta-Val, Betamethasone 17-valerate, Betamethasone valerate, Betnesol V, Betneval, Betnovate, Betnovateat, Betoid, Bettamousse, Bextasol, Celestan V, Celestane V, Celestoderm, Celestoderm V, Celeston valerate, Dermosol, Dermovaleas, Ecoval 70, Fuciderm, Hormezon, Rinderon V, Stanoval, Tokuderm, Valisone, (11beta,16beta)-9-Fluoro-11,21-dihydroxy-16-methyl-17-[(1-oxopentyl)oxy]pregna-1,4-diene-3,20-dione, 9-Fluoro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione 17-valerate, Valeric acid 17-ester with 9-fluoro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione, 9alpha-Fluoro-11beta,21-dihydroxy-16beta-methyl-17alpha-valeryloxypregna-1,4-diene-3,20-dione, 9alpha-Fluoro-16beta-methylprednisolone 17-valerate, Betamethasone 17alpha-valerate, beta-Methasone 17-valerate, CS valerate, Betamethasone-21-valerate |
| InChI Formula | InChI=1S/C27H37FO6/c1-5-6-7-23(33)34-27(22(32)15-29)16(2)12-20-19-9-8-17-13-18(30)10-11-24(17,3)26(19,28)21(31)14-25(20,27)4/h10-11,13,16,19-21,29,31H,5-9,12,14-15H2,1-4H3/t16-,19-,20-,21-,24-,25-,26-,27-/m0/s1 |
| SMILES | CCCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO |
| IUPAC Name | [(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] pentanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction