missing translation for 'onlineSavingsMsg'
Learn More

Betamethasone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30311350
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311350 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311350 Supplier Dr. Ehrenstorfer Supplier No. DREA10575000AL100
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Betamethasone
CAS 378-44-9
Analyte or Component Names Betamethasone
Shipping Condition Room Temperature
Molecular Formula C22 H29 F O5
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Betamethasone Acetate Imp. A (EP), Betamethasone Valerate Imp. A (EP), Dexamethasone Imp. B (EP), Betamethasone, 9-Fluoro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione, Betamethasone Dipropionate Imp. A (Pharmeuropa), CCI 922, Ph Eur Betamethasone Valerate Impurity A, Betamethasone Sodium Phosphate related, CS valerate, Betamethasone, Betamethasone Acetate Impurity A, Betamethasone Dipropionate Impurity A, Betamethasone Valerate Impurity A, Dexamethasone Impurity B
InChI Formula InChI=1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,17-,19-,20-,21-,22-/m0/s1
SMILES C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO
IUPAC Name (8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 392.46
Quantity 1 mL
Formula Weight 392.1999 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.