missing translation for 'onlineSavingsMsg'
Learn More

Betaine Hydrochloride, TRC

Product Code. 30138242
Change view
Click to view available options
Quantity:
2 mg
4 mg
20 mg
Unit Size:
20mg
2mg
4mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138242 4 mg 4mg
30135294 20 mg 20mg
17981135 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138242 Supplier Toronto Research Chemicals Supplier No. TRCB3251554MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-46-5
Molecular Formula C5 H12 N O2 . Cl
Molecular Weight (g/mol) 153.61
Synonym Betaine Hydrochloride, (Carboxymethyl)trimethylammonium chloride, 1-Carboxy-N,N,N-trimethylmethanaminium chloride, Achylin, Acidin, Acidine, Acidogeno, Acidol, Acidol hydrochloride, Acinorm, Acipepsol, Aciventral forte, Betaine chloride, Betainium hydrochloride, Glycine betaine hydrochloride, Glycocoll betaine hydrochloride, Pluchine, Rubrine C hydrochloride
IUPAC Name carboxymethyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CC(=O)O

Specifications

Analyte or Component Names Betaine hydrochloride
Shipping Condition Room Temperature
InChI Formula InChI=1S/C5H11NO2.ClH/c1-6(2,3)4-5(7)8;/h4H2,1-3H3;1H
Solution Type Neat
Industry Type Pharmaceutical
Quantity 4 mg
Formula Weight 153.0557
Chemical Name or Material Betaine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.