missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betaine Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Betaine hydrochloride |
| CAS | 590-46-5 |
| Analyte or Component Names | Betaine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H12 N O2 . Cl |
| Certifications/Compliance | ISO 9001 |
| Synonym | Betaine Hydrochloride, (Carboxymethyl)trimethylammonium chloride, 1-Carboxy-N,N,N-trimethylmethanaminium chloride, Achylin, Acidin, Acidine, Acidogeno, Acidol, Acidol hydrochloride, Acinorm, Acipepsol, Aciventral forte, Betaine chloride, Betainium hydrochloride, Glycine betaine hydrochloride, Glycocoll betaine hydrochloride, Pluchine, Rubrine C hydrochloride |
| InChI Formula | InChI=1S/C5H11NO2.ClH/c1-6(2,3)4-5(7)8;/h4H2,1-3H3;1H |
| Solution Type | Neat |
| SMILES | [Cl-].C[N+](C)(C)CC(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?