missing translation for 'onlineSavingsMsg'
Learn More

Betaine hydrochloride, 99%

Product Code. 11453157
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453157 1000 g 1000g
11463157 250 g 250g
11473157 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453157 Supplier Thermo Scientific Alfa Aesar Supplier No. A16122.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 590-46-5 | C5H12ClNO2 | 153.61 g/mol

Betaine hydrochloride is a naturally occurring compound that has been of interest for its role in osmoregulation. As a drug, it has been used as a source of hydrochloric acid in the treatment of hypochlorhydria. Betaine has also been used in the treatment of liver disorders, for hyperkalemia, for homocystinuria and for gastrointestinal disturbances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Betaine hydrochloride is a naturally occurring compound that has been of interest for its role in osmoregulation. As a drug, it has been used as a source of hydrochloric acid in the treatment of hypochlorhydria. Betaine has also been used in the treatment of liver disorders, for hyperkalemia, for homocystinuria and for gastrointestinal disturbances.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-46-5
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00011903
InChI Key HOPSCVCBEOCPJZ-UHFFFAOYSA-N
Synonym betaine hydrochloride, betaine chloride, pluchine, acidogeno, acipepsol, achylin, acidin, acinorm, acidol, acidine
PubChem CID 11545
SMILES [Cl-].C[N+](C)(C)CC(O)=O

Specifications

Melting Point ∼240°C (decomposition)
Quantity 1000 g
Beilstein 3916181
Merck Index 14,1179
Solubility Information Soluble in water.
IUPAC Name (carboxymethyl)trimethylazanium chloride
Formula Weight 153.61
Percent Purity 99%
Chemical Name or Material Betaine hydrochloride
compliance-icons
Product Identifier
  • Betaine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Betaine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.