Learn More
Betaine hydrochloride, 99%, extra pure, specified according to the requirements of U, Thermo Scientific Chemicals
£23.70 - £163.00
Chemical Identifiers
CAS | 590-46-5 |
---|---|
Molecular Formula | C5H12ClNO2 |
Molecular Weight (g/mol) | 153.61 |
MDL Number | MFCD00011903 |
InChI Key | HOPSCVCBEOCPJZ-UHFFFAOYSA-N |
Synonym | betaine hydrochloride, betaine chloride, pluchine, acidogeno, acipepsol, achylin, acidin, acinorm, acidol, acidine |
PubChem CID | 11545 |
IUPAC Name | carboxymethyl(trimethyl)azanium;chloride |
SMILES | [Cl-].C[N+](C)(C)CC(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10423405
|
Thermo Scientific Acros
164552500 |
250 g | Plastic bottle |
£23.70
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10783541
|
Thermo Scientific Acros
164550010 |
1 kg | Plastic bottle |
£67.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10117793
|
Thermo Scientific
164550025 |
2.5 kg | Plastic bottle |
£163.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
590-46-5 | |
153.61 | |
HOPSCVCBEOCPJZ-UHFFFAOYSA-N | |
11545 | |
[Cl-].C[N+](C)(C)CC(O)=O |
C5H12ClNO2 | |
MFCD00011903 | |
betaine hydrochloride, betaine chloride, pluchine, acidogeno, acipepsol, achylin, acidin, acinorm, acidol, acidine | |
carboxymethyl(trimethyl)azanium;chloride |
Specifications
590-46-5 | |
100.0 | |
Colorless to White | |
0.1% max. | |
99% | |
(CH3)3NCH2CO2H·Cl | |
15, 1181 | |
Solubility in water: 647g/L (25°C). Other solubilities: soluble in ethanol,practically insoluble in chloroform and ether | |
[Cl-].C[N+](C)(C)CC(O)=O | |
153.61 | |
153.61 | |
Extra Pure | |
βine hydrochloride |
98.5 | |
241.0°C | |
0.8 to 1.2 (1:4) | |
Authentic | |
C5H12ClNO2 | |
MFCD00011903 | |
betaine hydrochloride, betaine chloride, pluchine, acidogeno, acipepsol, achylin, acidin, acinorm, acidol, acidine | |
HOPSCVCBEOCPJZ-UHFFFAOYSA-N | |
carboxymethyl(trimethyl)azanium;chloride | |
11545 | |
99% | |
Crystals or Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-683-1
- Betaine hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only