missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betaine Ethyl Ester Chloride, Mikromol™
Description
Betaine Ethyl Ester Chloride
Specifications
Specifications
| Chemical Name or Material | Betaine Ethyl Ester Chloride |
| CAS | 11-9-3032 |
| Analyte or Component Names | Betaine Ethyl Ester Chloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H16 N O2 . Cl |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-Ethoxy-N,N,N-trimethyl-2-oxo-ethanaminium Chloride (1:1), (Carboxymethyl)trimethylammonium chloride, ethyl ester (7CI), Ammonium, (carboxymethyl)trimethyl-, chloride, ethyl ester (8CI), Ethanaminium, 2-ethoxy-N,N,N-trimethyl-2-oxo-, chloride (9CI), (Carbethoxymethyl)trimethylammonium chloride, Ethoxycarbonylmethyltrimethylammonium chloride |
| InChI Formula | InChI=1S/C7H16NO2.ClH/c1-5-10-7(9)6-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1 |
| SMILES | [Cl-].CCOC(=O)C[N+](C)(C)C |
| IUPAC Name | (2-ethoxy-2-oxoethyl)-trimethylazanium;chloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?