missing translation for 'onlineSavingsMsg'
Learn More

Betaine, anhydrous, 98%

Product Code. 11415847
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11415847

Brand: Thermo Scientific Alfa Aesar B24397.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-43-7 | C5H11NO2 | 117.148 g/mol

Betaine is an active ingredient in toothpaste to control the symptoms of dryness of the mouth. It is used to treat homocystinuria, which is a defect in the major pathway of methionine biosynthesis. It is also used for boosting the immune system and for improving athletic performance. It is helpful to prevent noncancerous tumors in the colon (colorectal adenomas).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Betaine is an active ingredient in toothpaste to control the symptoms of dryness of the mouth. It is used to treat homocystinuria, which is a defect in the major pathway of methionine biosynthesis. It is also used for boosting the immune system and for improving athletic performance. It is helpful to prevent noncancerous tumors in the colon (colorectal adenomas).

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-43-7
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00012123
InChI Key KWIUHFFTVRNATP-UHFFFAOYSA-N
Synonym betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin
PubChem CID 247
ChEBI CHEBI:17750
IUPAC Name 2-(trimethylazaniumyl)acetate
SMILES C[N+](C)(C)CC(=O)[O-]

Specifications

Melting Point >300°C (decomposition)
Assay Percent Range 0.98
Quantity 500 g
Beilstein 3537113
Sensitivity Hygroscopic
Merck Index 14,1179
Solubility Information Soluble in water and methanol.
Formula Weight 117.15
Percent Purity 98%
Chemical Name or Material Betaine, Anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.