missing translation for 'onlineSavingsMsg'
Learn More

Betahistine Mesylate, TRC

Product Code. 30340036
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340036 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340036 Supplier Toronto Research Chemicals Supplier No. TRCB324003500MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Betahistine mesilate
CAS 54856-23-4
Molecular Formula C8 H12 N2 . 2 C H4 O3 S
Quantity 500 mg
Synonym Betahistine mesilate, N-Methyl-2-(pyridin-2-yl)ethanamine bis(methanesulfonate), 2-Pyridineethanamine, N-methyl-, methanesulfonate (1:2), 2-Pyridineethanamine, N-methyl-, dimethanesulfonate (9CI), Betahistine mesylate, Merislon
InChI Formula InChI=1S/C8H12N2.2CH4O3S/c1-9-7-5-8-4-2-3-6-10-8;2*1-5(2,3)4/h2-4,6,9H,5,7H2,1H3;2*1H3,(H,2,3,4)
SMILES CNCCc1ccccn1.CS(=O)(=O)O.CS(=O)(=O)O
IUPAC Name methanesulfonic acid;N-methyl-2-pyridin-2-ylethanamine
Molecular Weight (g/mol) 328.41
Formula Weight 328.0763
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.