missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betahistine Mesilate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Betahistine Mesilate
Specifications
Specifications
| Chemical Name or Material | Betahistine mesilate |
| CAS | 54856-23-4 |
| Analyte or Component Names | Betahistine mesilate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H12 N2 . 2 C H4 O3 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | Betahistine mesilate, N-Methyl-2-(pyridin-2-yl)ethanamine bis(methanesulfonate), 2-Pyridineethanamine, N-methyl-, methanesulfonate (1:2), 2-Pyridineethanamine, N-methyl-, dimethanesulfonate (9CI), Betahistine mesylate, Merislon |
| InChI Formula | InChI=1S/C8H12N2.2CH4O3S/c1-9-7-5-8-4-2-3-6-10-8;2*1-5(2,3)4/h2-4,6,9H,5,7H2,1H3;2*1H3,(H,2,3,4) |
| SMILES | CNCCc1ccccn1.CS(=O)(=O)O.CS(=O)(=O)O |
| IUPAC Name | methanesulfonic acid;N-methyl-2-pyridin-2-ylethanamine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction