missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betahistine Dihydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Betahistine Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Betahistine dihydrochloride |
| CAS | 5579-84-0 |
| Analyte or Component Names | Betahistine dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H12 N2 . 2 Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Betahistine Dihydrochloride, N-Methyl-2-pyridineethanamine hydrochloride (1:2), N-Methyl-2-pyridineethanamine dihydrochloride, 2-[2-(Methylamino)ethyl]pyridine dihydrochloride, Betahistine dihydrochloride, Betahistine hydrochloride, Betaserc, PT 9, Serc, Sinmenier, Vasomotal |
| InChI Formula | InChI=1S/C8H12N2.2ClH/c1-9-7-5-8-4-2-3-6-10-8;;/h2-4,6,9H,5,7H2,1H3;2*1H |
| SMILES | Cl.Cl.CNCCc1ccccn1 |
| IUPAC Name | N-methyl-2-pyridin-2-ylethanamine;dihydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction