missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Order Info
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
Specifications
| Column Format | Analytical Column |
| Diameter (Metric) | 4.6 mm |
| Film Thickness | 5 μm |
| For Use With | LC/MS Separation |
| Functionality/Interaction Type | Stable bonding |
| Length (Metric) | 250 mm |
| Particle Shape | Spherical |
| Particle Size | 5 μm |
| Pore Size | 150 Å |
| Surface Area | 200 m²/g |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?