Learn More
Thermo Scientific™ BetaBasic™ CN HPLC Columns, 5μm Particle Size
Thermo ScientificTM BetaBasicTM CN HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
Brand: Thermo Scientific™ 71705-154630
Code : 16
Additional Details : Weight : 0.10000kg
Description
Thermo ScientificTM BetaBasicTM CN HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
- Pore size suitable for small molecules, peptides and protein digests
- Superb pH stability
Ordering Information:
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
Analytical Column | |
LC/MS, APCI | |
5 μm | |
150 Å | |
L10 | |
Beta | |
5% | |
Yes |
4.6 mm | |
150 mm | |
2 to 9 | |
200 m²/g | |
Cyano (CN) | |
Silica | |
Mixed Mode | |
Normal and Reversed Phase |