Learn More
Thermo Scientific™ BetaBasic™ 18 HPLC Columns, 5μm Particle Size
Available in limited dimensions. Please contact Customer Service for more details.
Brand: Thermo Scientific™ 71505-152130
Code : 16
Additional Details : Weight : 0.10000kg
Description
Thermo ScientificTM BetaBasicTM 18 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
- Pore size suitable for small molecules, peptides and protein digests
- Superb pH stability
Ordering Information:
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
Analytical Column | |
LC/MS, APCI | |
5 μm | |
150 Å | |
L1 | |
Beta | |
13% | |
Yes |
2.1 mm | |
150 mm | |
2 to 12 | |
200 m²/g | |
C18 | |
Silica | |
Reversed Phase | |
Reversed Phase |