Learn More
Thermo Scientific™ BetaBasic™ 18 Guard Cartridges
Available in limited dimensions. Please contact Customer Service for more details.
Brand: Thermo Scientific™ 71503-013001
Code : 16
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific™ BetaBasic™ 18 Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.
- The 10mm design offers maximum protection with minimal increase in retention
- Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
- Highly efficient and reproducible Thermo Scientific columns
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Requires:
Thermo Scientific Unigard™ direct connection or stand-alone holder, sold separately.
Specifications
13% | |
3 μm | |
150Å | |
3 mm | |
Yes |
2 to 12 | |
10 mm | |
200 m2/g | |
Uniguard Direct-connection and Stand Alone Holder | |
L1 |