missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta ureidopropionase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35555-20ul
This item is not returnable.
View return policy
Description
beta ureidopropionase Polyclonal antibody specifically detects beta ureidopropionase in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| beta ureidopropionase | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| Beta-alanine synthase, beta-ureidopropionase, BUP-1, BUP1EC 3.5.1.6, N-carbamoyl-beta-alanine amidohydrolase, ureidopropionase, beta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human beta ureidopropionase (NP_057411.1).,, Sequence:, MAGAEWKSLEECLEKHLPLPDLQEVKRVLYGKELRKLDLPREAFEAASREDFELQGYAFEAAEEQLRRPRIVHVGLVQNRIPLPANAPVAEQVSALHRRIKAIVEVAAMCGVNIICFQEAWTMPFAFCTREKLPWTEFAESAEDGPTTRFCQKLAKNHDMVVVSPILERDSEHGDVLWNTAVVISNSGAVLGKTRKNHIPRVGDFNESTYYMEGNLGHPVFQTQFGRIAVNICYGRHHPL | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51733 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction