missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Tubulin, Rabbit anti-Human, Clone: S11B, Novus Biologicals™
Rabbit Monoclonal Antibody
£217.00 - £463.00
Specifications
| Antigen | beta Tubulin |
|---|---|
| Clone | S11B |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18328010
|
Novus Biologicals
NBP2-81064-0.025MG |
0.025 mg |
£217.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18384100
|
Novus Biologicals
NBP2-81064-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta Tubulin Monoclonal specifically detects beta Tubulin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| beta Tubulin | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| PBS | |
| 203068 | |
| Human MBP (microtubule-binding protein). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S11B | |
| Western Blot, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta | |
| TUBB | |
| IgG λ | |
| Protein A purified | |
| Binds specifically to human beta Tubulin (no crossreactivity with alpha-tubulin). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title