missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-Tubulin III Mouse, anti-Human, Unconjugated, Clone: AA10, STEMCELL Technologies
Mouse Monoclonal Antibody
£155.25 - £375.30
Specifications
| Antigen | Beta-Tubulin III |
|---|---|
| Clone | AA10 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17109011
|
Stemcell Technologies
60100.1 |
25 μg |
£155.25
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17199001
|
Stemcell Technologies
60100 |
100 μg |
£375.30
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The AA10 antibody reacts with beta-tubulin III, an ∼50 - 55 kDa structural protein that is a constituent of tubulin
- Tubulin is the major component of microtubules within the cytoskeleton and is assembled from heterodimers of alpha and beta tubulin subunits
- The beta III isoform of tubulin, also known as neuron-specific class III beta-tubulin, is expressed primarily in neurons and is widely used as a marker to distinguish neurons from other cell types
- Beta-tubulin III contributes to microtubule formation in neuronal cell bodies and axons, a function involving GTP binding, and plays roles in axonal transport, neuronal cell proliferation, and differentiation
- It is highly expressed in several types of cancer and is a predictive and prognostic marker for various tumors, for example, being found in neoplastic but not in normal glial cells
- The AA10 antibody is expected to recognize all mammalian homologs of beta-tubulin III
Specifications
| Beta-Tubulin III | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Human, Rat, Mouse | |
| Primary |
| AA10 | |
| Monoclonal | |
| Mouse | |
| IgG2a κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title