missing translation for 'onlineSavingsMsg'
Learn More

beta Tubulin Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18405294
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405294 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405294 Supplier Novus Biologicals (Bio-Techne) Supplier No. NB600936MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

beta Tubulin Polyclonal antibody specifically detects beta Tubulin in Human, Mouse, Rat, Porcine, Bovine, C. elegans, Chicken, Chinese Hamster, Insect, Invertebrate, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Tubulin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta
Host Species Rabbit
Immunogen A synthetic peptide made to the N-terminal region of human beta Tubulin (within residues 1-100). [Swiss-Prot: P07437]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cell Biology, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 203068
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.