missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Tubulin 2 Polyclonal Antibody

beta Tubulin 2 Polyclonal Antibody
Brand: Invitrogen PA5107300
This item is not returnable.
View return policy
Description
beta Tubulin 2 Polyclonal Antibody for Western Blot
Specifications
| beta Tubulin 2 | |
| Polyclonal | |
| Unconjugated | |
| TUBB2A | |
| 2410129E14Rik; 2t; b2 tubulin; B2t; bA506K6.1; beta 2 tubulin; BETA 85D; beta tubulin; beta(2)Tu; beta(2)Tub; beta(2)Tubulin; beta;Tub85D; beta[[2]]-tubulin; BETA2; beta2 tubulin; beta2t; beta2tub; beta2-tub; beta2tubulin; Beta-2-tubulin; beta2-tubulin; beta85D; betaTub; beta-tub; betaTub2; BetaTub85D; beta-tub85D; betaTub85D-PA; beta-Tubulin; beta-Tubulin 85D; beta-Tubulin at 85D; beta-tubulin T beta15 (aa 1-445); betaTubulin85D; beta-tubulin85D; brdp; CDCBM5; CG9359; CG9359-PA; class II beta-tubulin isotype; class IIa beta-tubulin; class IIb beta-tubulin; D.m.BETA-85D; dJ40E16.7; Dmbeta2; Dmel\CG9359; Dmel_CG9359; DTB4; GH02051p; M(beta)2; ms(3)KK[D]; PMGYSA; RGD1309427; t beta-15; testis-specific beta-tubulin; Tub; Tub2A; TUBB; TUBB2; TUBB2A; tubb2a {ECO:0000250; Tubb2b; TubB85D; tubulin; tubulin beta; tubulin beta 2A class IIa; tubulin beta 2B class IIb; Tubulin beta class IIa; Tubulin beta-2 chain; Tubulin beta-2A chain; tubulin beta-2B chain; tubulin, beta 2; tubulin, beta 2A class IIa; tubulin, beta 2B class IIb; tubulin, beta polypeptide; tubulin, beta polypeptide 2; tubulin, beta polypeptide paralog; UniProtKB:Q7TMM9} | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 347733, 7280 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q13885, Q9BVA1 | |
| TUBB2A, TUBB2B | |
| Synthetic peptide directed towards the N-terminal region of Human TUBB2B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction