missing translation for 'onlineSavingsMsg'
Learn More

beta-TrCP1/BTRC Antibody (OTI2H2), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18422087 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422087 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422087 Supplier Novus Biologicals Supplier No. NBP271402MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-TrCP1/BTRC Monoclonal antibody specifically detects beta-TrCP1/BTRC in Human, Mouse, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-TrCP1/BTRC
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2H2
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias beta-transducin repeat containing, betaTrCP, BETA-TRCP, beta-TrCP1, bTrCP, bTrCP1, E3RSIkappaB, F-box and WD repeats protein beta-TrCP, F-box/WD repeat-containing protein 1A, FBW1A, FBXW1, FBXW1ABTRCP, Fwd1, MGC4643, pIkappaBalpha-E3 receptor subunit
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 52-354 of human BTRC(NP_378663) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 8945
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.