missing translation for 'onlineSavingsMsg'
Learn More

beta-Synuclein Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18402459 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402459 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402459 Supplier Novus Biologicals Supplier No. NBP305529AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-Synuclein Monoclonal antibody specifically detects beta-Synuclein in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-Synuclein
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias beta-synuclein, synuclein, beta
Host Species Mouse
Immunogen C-terminal peptide of human beta-Synuclein EPEGESYEDPPQEEYQEYEPEA coupled to KLH
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6620
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.