missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Sarcoglycan Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38356-100ul
This item is not returnable.
View return policy
Description
beta Sarcoglycan Polyclonal antibody specifically detects beta Sarcoglycan in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| beta Sarcoglycan | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| 43 kDa dystrophin-associated glycoprotein, 43DAG, A3bbeta-SG, beta-sarcoglycan, beta-sarcoglycan(43kD dystrophin-associated glycoprotein), Beta-SG, LGMD2E, limb girdle muscular dystrophy 2E (non-linked families), sarcoglycan, beta (43kD dystrophin-associated glycoprotein), sarcoglycan, beta (43kDa dystrophin-associated glycoprotein), SGC | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 94-318 of human beta Sarcoglycan (NP_000223.1).,, Sequence:, PNGCDSMEFHESGLLRFKQVSDMGVIHPLYKSTVGGRRNENLVITGNNQPIVFQQGTTKLSVENNKTSITSDIGMQFFDPRTQNILFSTDYETHEFHLPSGVKSLNVQKASTERITSNATSDLNIKVDGRAIVRGNEGVFIMGKTIEFHMGGNMELKAENSIILNGSVMVSTTRLPSSSSGDQLGSGDWVRYKLCMCADGTLFKVQVTSQNMGCQISDNPCGNTH | |
| 100 μL | |
| Signal Transduction | |
| 6443 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction