missing translation for 'onlineSavingsMsg'
Learn More

Beta-Pseudouridine, TRC

Product Code. 30135291
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135291 2.5 mg 2.5mg
30006038 25 mg 25mg
30005595 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135291 Supplier Toronto Research Chemicals Supplier No. TRCP8396052.5MG
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1445-07-4
Synonym 5-β-D-Ribofuranosyl-2,4(1H,3H)-Pyrimidinedione∣5-(β-D-Ribofuranosyl)uracil∣5-Ribosyluracil∣NSC 162405∣Pseudouridine C∣β-D-Pseudouridine∣Pseudouridine∣ψ-Uridine;

Specifications

Format Neat
Molecular Weight (g/mol) 244.2
Shipping Condition Room Temperature
Quantity 2.5 mg
Chemical Name or Material beta-Pseudouridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.