missing translation for 'onlineSavingsMsg'
Learn More

beta-Nicotinamide adenine dinucleotide phosphate monosodium salt, Thermo Scientific Chemicals

Catalog Number 11349608
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1184-16-3
C21H27N7NaO17P3
765.39
MFCD00036973
JNUMDLCHLVUHFS-QYZPTAICSA-M
nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium
131845262
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
This item is not returnable. View return policy
1184-16-3
C21H27N7NaO17P3
765.39
MFCD00036973
JNUMDLCHLVUHFS-QYZPTAICSA-M
nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium
131845262
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

β-Nicotinamide adenine dinucleotide phosphate monosodium salt acts as an electron acceptor used by cells. It is also useful in vitro to study NADP. Furthermore, it is used in a wide range of enzyme catalyzed oxidation reduction reactions. The combination of NADP+/NADPH is employed in a variety of antioxidation mechanisms, which protect against reactive oxidation species accumulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 1 g
Beilstein 4779954
Solubility Information Soluble in water.
Formula Weight 765.4
Physical Form Powder
Chemical Name or Material beta-Nicotinamide adenine dinucleotide phosphate monosodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'