missing translation for 'onlineSavingsMsg'
Learn More

beta-Nicotinamide adenine dinucleotide, disodium salt, hydrate, 93+%, reduced form

Product Code. 10207713 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10207713 5 g Glass bottle 5g
10184630 250 mg Glass bottle 250mg
10711911 1 g Glass bottle 1g
10034203 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10207713 Supplier Thermo Scientific Acros Supplier No. 271100050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1949720-50-6 | C21H27N7Na2O14P2 | 709.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1949720-50-6
Molecular Formula C21H27N7Na2O14P2
Molecular Weight (g/mol) 709.41
MDL Number MFCD00036200
InChI Key QRGNQKGQENGQSE-WUEGHLCSSA-L
Synonym beta-nadh disodium salt
PubChem CID 131673989
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium
SMILES [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Absorbance 0.50mg/mL NADH in 10mM TRIS solution
CAS Min % 95.0
CAS Max % 100.0
Melting Point 140.0°C to 142.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 709.4
Percent Purity 95+%
Physical Form Powder
Chemical Name or Material β-Nicotinamide adenine dinucleotide, disodium salt, hydrate, Reduced Form
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.