missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ beta-NGF Antibody (fasinumab), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28872-100ug
This item is not returnable.
View return policy
Description
beta-NGF Monoclonal antibody specifically detects beta-NGF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| beta-NGF | |
| Monoclonal | |
| Unconjugated | |
| P01138 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| fasinumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428 | |
| NGF/ bNGF | |
| 100 μg | |
| Cancer, Neuroscience | |
| 4803 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction