missing translation for 'onlineSavingsMsg'
Learn More

beta-NGF Antibody (25623) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30734982 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734982 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734982 Supplier Novus Biologicals Supplier No. FAB256I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-NGF Monoclonal antibody specifically detects beta-NGF in Human samples. It is validated for ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-NGF
Applications ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 25623
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human β-NGF, Ser122-Ala241, Accession # CAA36832
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4803
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.