missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
beta-NGF Monoclonal antibody specifically detects beta-NGF in Mouse samples. It is validated for ELISA
Specifications
Specifications
| Antigen | beta-NGF |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2529A |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428 |
| Host Species | Rabbit |
| Immunogen | Mouse myeloma cell line, NS0-derived mouse β-NGF, Ser122-Gly241, Accession # Q6LDU8 |
| Purification Method | Protein A or G purified from cell culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?