missing translation for 'onlineSavingsMsg'
Learn More

beta-Naphthol Violet, Thermo Scientific™

Product Code. 11475687
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11475687

Brand: Thermo Scientific Alfa Aesar B24279.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Beta-Naphthol violet exhibits antiseptic properties and is used in the synthesis of certain azo dyes and antioxidants for rubber. It can also be used in the detection of the ferric ion which turns the Beta-Naphthol violet solution green.

Solubility
Soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7143-21-7
Molecular Formula C16H9N3Na2O9S2
Molecular Weight (g/mol) 497.36
MDL Number MFCD00021457
InChI Key QMNPQXAVQXMXTD-UHFFFAOYSA-L
PubChem CID 44135658
IUPAC Name disodium;4-[(4-nitrophenyl)hydrazinylidene]-3-oxonaphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1NN=C2C3=C(C=C(C=C3)S(=O)(=O)[O-])C=C(C2=O)S(=O)(=O)[O-])[N+](=O)[O-].[Na+].[Na+]

Specifications

Solubility Information Soluble in water.
Formula Weight 497.37
Odor Odorless
Quantity 250 g
Chemical Name or Material beta-Naphthol Violet
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.