missing translation for 'onlineSavingsMsg'
Learn More

Beta-Naphthoflavone, TRC

Product Code. 30480115
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30480115 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30480115 Supplier Toronto Research Chemicals Supplier No. TRCN369015100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6051-87-2
Molecular Formula C19 H12 O2
Quantity 100 mg
Synonym 1H-Naphtho[2,1-b]pyran-1-one, 3-phenyl-, 5,6-Benzoflavone (6CI,7CI), 3-Phenyl-1H-naphtho[2,1-b]pyran-1-one, 3-Phenyl-1H-benzo[f]chromen-1-one, BNF, NSC 136015, β-Naphthoflavone
InChI Formula InChI=1S/C19H12O2/c20-16-12-18(14-7-2-1-3-8-14)21-17-11-10-13-6-4-5-9-15(13)19(16)17/h1-12H
SMILES O=C1C=C(Oc2ccc3ccccc3c12)c4ccccc4
IUPAC Name 3-phenylbenzo[f]chromen-1-one
Molecular Weight (g/mol) 272.3
Formula Weight 272.0837
Chemical Name or Material beta-Naphthoflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.