missing translation for 'onlineSavingsMsg'
Learn More

beta-Naphthoflavone, 98+%

Product Code. 11458467
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458467 1 g 1g
11468467 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458467 Supplier Thermo Scientific Alfa Aesar Supplier No. A18543.03
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6051-87-2 | C19H12O2 | 272.303 g/mol

Inhibitor of aryl hydrocarbon hydroxylase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6051-87-2
Molecular Formula C19H12O2
Molecular Weight (g/mol) 272.303
MDL Number MFCD00004986
InChI Key OUGIDAPQYNCXRA-UHFFFAOYSA-N
Synonym beta-naphthoflavone, 5,6-benzoflavone, beta-nf, 3-phenyl-1h-naphtho 2,1-b pyran-1-one, 3-phenyl-1h-benzo f chromen-1-one, 1h-naphtho 2,1-b pyran-1-one, 3-phenyl, unii-1bt0256y8o, 3-phenylbenzo f chromen-1-one, ccris 3262, .beta.-naphthoflavone
PubChem CID 2361
ChEBI CHEBI:77013
IUPAC Name 3-phenylbenzo[f]chromen-1-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC4=CC=CC=C43

Specifications

Melting Point 162°C to 166°C
Odor Odorless
Quantity 1 g
Beilstein 18991
Formula Weight 272.31
Percent Purity ≥98%
Chemical Name or Material beta-Naphthoflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.