missing translation for 'onlineSavingsMsg'
Learn More

beta-NADH Disodium Salt, TRC

Product Code. 30465424
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465424 1 g 1g
30469571 500 mg -
30484131 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30465424 Supplier Toronto Research Chemicals Supplier No. TRCN2014801G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-68-8
Molecular Formula C21 H27 N7 O14 P2 . 2 Na
Molecular Weight (g/mol) 709.4
Synonym Adenosine 5'-(trihydrogen diphosphate), P'→5'-ester with 1,4-dihydro-1-β-D-ribofuranosylnicotinamide, sodium salt (1:2), Adenosine 5'-(trihydrogen diphosphate), P'→5'-ester with 1,4-dihydro-1-β-D-ribofuranosyl-3-pyridinecarboxamide, disodium salt (9CI), Adenosine 5'-(trihydrogen pyrophosphate), 5'→5'-ester with 1,4-dihydro-1-β-D-ribofuranosylnicotinamide, disodium salt (8CI), Disodium NADH, NADH disodium salt, Reduced nicotinamide adenine diphosphate disodium salt, Reduced nicotinamide-adenine dinucleotide disodium salt, Reduced nicotinamide-adenine dinucleotide (NADH) disodium salt, β-DPNH disodium salt, β-NADH disodium salt, β-Nicotinamide adenine dinucleotide reduced disodium salt
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]2O[C@H](COP(=O)([O-])OP(=O)([O-])OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H29N7O14P2.2Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;/h1,3-4,7-8,10-11,13-16,20-21,29-32H,2,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);;/q;2*+1/p-2/t10-,11-,13-,14-,15-,16-,20-,21-;;/m1../s1
Formula Weight 709.0887
Chemical Name or Material beta-NADH Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.