missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Beta Lactoglobulin Polyclonal antibody specifically detects Beta Lactoglobulin in Bovine samples. It is validated for Western Blot, ELISA, Functional Assay, Immunoelectrophoresis, Functional Assay
Specifications
Specifications
| Antigen | Beta Lactoglobulin |
| Applications | Western Blot, ELISA, Functional Assay, Immunoelectrophoresis, Functional Assay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Host Species | Rabbit |
| Immunogen | Beta lactoglobulin extract from bovine milk. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?