missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta Lactamase Monoclonal Antibody (3E11.G3), Invitrogen™

Mouse Monoclonal Antibody
£367.00 - £916.00
Specifications
| Antigen | beta Lactamase |
|---|---|
| Clone | 3E11.G3 |
| Concentration | 0.5 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17850146
|
Invitrogen™
MA5-46174 |
100 μg |
£367.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11588972
![]() |
Invitrogen™
MA110713 |
500 μg |
£465.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17840496
|
Invitrogen™
MA5-46121 |
1 mg |
£916.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MA1-10713 detects beta Lactamase from Enterobacter cloacae samples. MA1-10713 has been successfully used in ELISA applications. The MA1-10713 immunogen is Type IV beta-lactamase from Enterobacter cloacae .
The beta lactam antibiotics (penicillins and cephalosporins) are the most frequently used antimicrobial agents. All of the beta lactams are structurally related through the presence of a core beta lactam ring. Bacterial resistance to beta lactams continues to increase, primarily due to the production of beta lactamases. Beta lactamases catalyze the hydrolysis of the beta lactam bond, which destroys antibacterial activity. Bacteria that produce TEM type or SHV type beta lactamases have point mutations in structural genes that have extended the substrate specificity of these beta lactamases. As a result, many of the beta lactamase producing Gram negative pathogens have become multidrug resistant.Specifications
| beta Lactamase | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| Type IV beta-lactamase from Enterobacter cloacae. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles |
| 3E11.G3 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| blaCTX-M-14; FLJ14902; G24; lactamase beta; LACTB; mitochondrial 39S ribosomal protein L56; MRPL56; Serine beta-lactamase-like protein LACTB, mitochondrial; UNQ843/PRO1781 | |
| IgG3 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title