missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta L Crystallin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119054
This item is not returnable.
View return policy
Description
beta L Crystallin Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The uronate cycle functions as an alternative glucose metabolic pathway, accounting for about 5% of daily glucose catabolism. The product of this gene catalyzes the dehydrogenation of L-gulonate into dehydro-L-gulonate in the uronate cycle. The enzyme requires NAD(H) as a coenzyme, and is inhibited by inorganic phosphate. A similar gene in the rabbit is thought to serve a structural role in the lens of the eye.
Specifications
| beta L Crystallin | |
| Polyclonal | |
| Unconjugated | |
| CRYL1 | |
| 1110025H08Rik; A230106J09Rik; C85932; CRY; Cryl1; crystallin lambda 1; crystallin, lambda 1; crystallin, lamda 1; epididymis luminal protein 30; GDH; gul3dh; HEL30; lambda-CRY; Lambda-crystallin homolog; L-gulonate 3-dehydrogenase; testicular tissue protein Li 44 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 51084, 68631 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.59 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q99KP3, Q9Y2S2 | |
| CRYL1 | |
| Recombinant protein Crystallin Lambda 1. The antigen corresponds to amino acid range 24-232 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction